Merck
  • Novel unconditioned prosocial behavior in prairie voles (Microtus ochrogaster) as a model for empathy.
Novel unconditioned prosocial behavior in prairie voles (Microtus ochrogaster) as a model for empathy.
BMC research notes (2018-12-05)
Lucas A Stetzik, Alana W Sullivan, Heather B Patisaul, Bruce S Cushing
ABSTRACT
In this study, empathy is quantified using a novel social test. Empathy and prosocial behavior are linked to the expression of oxytocin in humans and rodent models. Specifically, prosocial behavior in prairie voles (Microtus ochrogaster) has been linked to the expression of oxytocin in the paraventricular nucleus of the hypothalamus. The animal's behavior was considered empathic if it spends significantly more time attempting to remove a loos fitting restraint (tether) from the stimulus animal than time in contact with a, simultaneously presented, non-social object similar to the tether. The behavioral data was cross-referenced with the number of neurons expressing oxytocin and arginine vasopressin, as well as the density of dopaminergic neurons (identified by the expression of tyrosine hydroxylase), in the paraventricular nucleus of the hypothalamus. These proteins influence empathic behavior in humans, non-human primates, rats, mice, and prairie voles. The consistency between neuroanatomical mechanisms linked to empathy, and the durations of time spent engaging in empathic contact, support the prediction that the empathic contact in this test is a distinct prosocial behavior, lacking prior behavioral training or the naturally occurring ethological relevance of other prosocial behaviors, and is a measure of empathy.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit