Merck
  • YB-1 alters MT1-MMP trafficking and stimulates MCF-7 breast tumor invasion and metastasis.
YB-1 alters MT1-MMP trafficking and stimulates MCF-7 breast tumor invasion and metastasis.
Biochemical and biophysical research communications (2010-07-06)
David H Lovett, Sunfa Cheng, Leslie Cape, Allan S Pollock, Peter R Mertens
ABSTRACT
YB-1 is a member of the cold shock domain family, with complex roles in DNA structure, gene transcription and translation. YB-1 promotes chromosomal instability, and mammary gland transgenic expression induces tumors with 100% penetrance. YB-1 is linked to poor prognosis in breast carcinoma and is a strong predictor of relapse and disease-specific survival. Survival is directly tied to the extent of local invasion and distal metastasis, processes dependent upon the activity of the membrane type I-matrix metalloproteinase, MT1-MMP. Non-invasive MCF-7 breast adenocarcinoma cells were transfected with YB-1/EGFP. YB-1 protein was detected in the invadopodia of cells with a migratory phenotype. There was increased expression of MT1-MMP protein concentrated at the leading edges of motile cells, which were highly invasive in collagen three-dimensional culture. The rates of MT1-MMP protein endocytosis and recycling to the cell surface were elevated in clones expressing higher levels of YB-1 protein. Control MCF-7 cells formed nonfatal, non-invasive, differentiated adenocarcinomas in vivo. MCF-7 cells expressing a twofold increase in YB-1 formed highly anaplastic tumors with local invasion, pulmonary metastases and high lethality. We conclude that YB-1 contributes to the development of an invasive, metastatic breast carcinoma phenotype by enhanced presentation of MT1-MMP at the sites of cellular invasion.
Sigma-Aldrich
Anti-MMP-14 Antibody, catalytic domain, clone 114-6G6, clone 114-6G6, Chemicon®, from mouse
Pricing and availability is not currently available.