- Identification and characterization of a Neospora caninum microneme-associated protein (NcMIC4) that exhibits unique lactose-binding properties.
Identification and characterization of a Neospora caninum microneme-associated protein (NcMIC4) that exhibits unique lactose-binding properties.
Microneme proteins have been shown to play an important role in the early phase of host cell adhesion, by mediating the contact between the parasite and host cell surface receptors. In this study we have identified and characterized a lectin-like protein of Neospora caninum tachyzoites which was purified by alpha-lactose-agarose affinity chromatography. Upon separation by sodium dodecyl sulfate-polyacrylamide gel electrophoresis, this lactose-binding protein migrated at 70 and 55 kDa under reducing and nonreducing conditions, respectively. Immunofluorescence and immunogold electron microscopy with affinity-purified antibodies showed that the protein was associated with the tachyzoite micronemes. Mass spectrometry analyses and expressed sequence tag database mining revealed that this protein is a member of the Neospora microneme protein family; the protein was named NcMIC4 (N. caninum microneme protein 4). Upon two-dimensional gel electrophoresis, NcMIC4 separated into seven distinct isoforms. Incubation of extracellular parasites at 37 degrees C resulted in the secretion of NcMIC4 into the medium as a soluble protein, and the secreted protein exhibited a slightly reduced M(r) but retained its lactose-binding properties. Immunofluorescence was used to investigate the temporal and spatial distribution of NcMIC4 in tachyzoites entering their host cells and showed that reexpression of NcMIC4 took place 30 min after entry into the host cell. Incubation of secreted fractions and purified NcMIC4 with Vero cells demonstrated binding of NcMIC4 to Vero cells as well as binding to chondroitin sulfate A glycosaminoglycans.