- Susceptibility of Pseudomonas pseudomallei to new beta-lactam and aminoglycoside antibiotics.
Susceptibility of Pseudomonas pseudomallei to new beta-lactam and aminoglycoside antibiotics.
The Journal of antimicrobial chemotherapy (1988-02-01)
D W McEniry, S H Gillespie, D Felmingham
PMID3162898
ABSTRACT
Pseudomonas pseudomallei is resistant to many antimicrobial agents. We determined the susceptibility to twelve beta-lactams and six aminoglycosides of twelve isolates of P. pseudomallei. Imipenem (MIC range 0.5-1.0 mg/l; MIC90 1.0 mg/l), ceftazidime (MIC range 1-8 mg/l; MIC90 8.0 mg/l), amoxycillin/clavulanate (MIC range 4-8 mg/l; MIC90 8.0 mg/l), piperacillin (MIC range 4-16 mg/l; MIC90 8.0 mg/l), and carumonam (MIC range 4-16 mg/l; MIC90 8.0 mg/l) were the most active. Aminoglycosides were relatively inactive, for example, gentamicin (MIC range 16-64 mg/l; MIC90 64 mg/l), netilmicin (MIC range 16-128 mg/l; MIC90 128 mg/l) and amikacin (MIC range 32-64 mg/l; MIC90 64 mg/l).
MATERIALS