Merck
  • Inhibition of Lats1/p-YAP1 pathway mitigates neuronal apoptosis and neurological deficits in a rat model of traumatic brain injury.
Inhibition of Lats1/p-YAP1 pathway mitigates neuronal apoptosis and neurological deficits in a rat model of traumatic brain injury.
CNS neuroscience & therapeutics (2018-03-01)
Di Li, Jia-Xuan Ji, Yi-Tian Xu, Hai-Bo Ni, Qin Rui, Hui-Xiang Liu, Feng Jiang, Rong Gao, Gang Chen
ABSTRACT
To investigate the roles of Lats1/p-YAP1 pathway in TBI-induced neuronal apoptosis and neurological deficits in rats. We found that Lats1 and YAP1 were expressed in cerebral cortex neurons of Sprague-Dawley rats, and the phosphorylation levels of Lats1 and YAP1 in injured regions were significantly increased after TBI. Furthermore, inhibition of Lats1 not only decreased the level of p-YAP1, but also attenuated neuronal apoptosis and neurological impairment. Our work demonstrates that inhibition of Lats1/p-YAP1 pathway mitigates neuronal apoptosis and neurological deficits in a rat model of TBI.
Sigma-Aldrich
Anti-LATS1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse