Merck
  • Mesenchymal stromal cell density is increased in higher grade myelodysplastic syndromes and independently predicts survival.
Mesenchymal stromal cell density is increased in higher grade myelodysplastic syndromes and independently predicts survival.
American journal of clinical pathology (2014-11-13)
Ryan C Johnson, Jason H Kurzer, Peter L Greenberg, Dita Gratzinger
ABSTRACT
We retrospectively tested the prognostic and diagnostic significance of CD271+ mesenchymal stromal cell (MSC) density in cytopenic patients who underwent bone marrow biopsy to evaluate for myelodysplastic syndromes (MDS). CD271+ MSC density was quantitated by automated image analysis of tissue microarray cores in 125 cytopenic patients: 40 lower grade MDS (<5% marrow blasts), 24 higher grade MDS, and 61 benign. CD271+ MSC density was increased in higher grade MDS compared with benign (P = .006) and lower grade MDS (P = .02). CD271+ MSC density was predictive of survival among patients with MDS independent of Revised International Prognostic Scoring System (IPSS-R), history of transfusion, therapy-related MDS, and fibrosis (hazard ratio, 3.4; P < .001). Among low or intermediate IPSS-R patients, median survival was significantly shorter in the high CD271+ MSC density group (47 vs 18 months, P < .02). High CD271+ MSC density is characteristic of higher grade MDS and is associated with poor risk independent of known prognostic factors.
Sigma-Aldrich
Anti-NGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.