Merck
  • Bioinspired Thiophosphorodichloridate Reagents for Chemoselective Histidine Bioconjugation.
Bioinspired Thiophosphorodichloridate Reagents for Chemoselective Histidine Bioconjugation.
Journal of the American Chemical Society (2019-04-25)
Shang Jia, Dan He, Christopher J Chang
ABSTRACT
Site-selective bioconjugation to native protein residues is a powerful tool for protein functionalization, with cysteine and lysine side chains being the most common points for attachment owing to their high nucleophilicity. We now report a strategy for histidine modification using thiophosphorodichloridate reagents that mimic post-translational histidine phosphorylation, enabling fast and selective labeling of protein histidines under mild conditions where various payloads can be introduced via copper-assisted alkyne-azide cycloaddition (CuAAC) chemistry. We establish that these reagents are particularly effective at covalent modification of His-tags, which are common motifs to facilitate protein purification, as illustrated by selective attachment of polyarginine cargoes to enhance the uptake of proteins into living cells. This work provides a starting point for probing and enhancing protein function using histidine-directed chemistry.
Sigma-Aldrich
Thiophosphoro alkyne dichloridate, ≥95%
Pricing and availability is not currently available.