- Peptide Bioink: Self-Assembling Nanofibrous Scaffolds for Three-Dimensional Organotypic Cultures.
Peptide Bioink: Self-Assembling Nanofibrous Scaffolds for Three-Dimensional Organotypic Cultures.
Nano letters (2015-07-28)
Yihua Loo, Anupama Lakshmanan, Ming Ni, Lai Ling Toh, Shi Wang, Charlotte A E Hauser
PMID26214046
ABSTRACT
Printable scaffolds with adequate mechanical strength and stiffness are sought after to ensure viability of printed cells and tissues. We report the first peptide bioinks-lysine-containing hexapeptides that self-assemble into stable, nanofibrous three-dimensional hydrogels with unprecedented stiffness of up to 40 kPa. These biocompatible scaffolds support the three-dimensional culture of human stem cells and differentiation of primary cells into organotypic (gastrointestinal and skin) structures for high-throughput screening, diagnosis, and tissue engineering.