Merck
  • Sphingosine-1-phosphate promotes erythrocyte glycolysis and oxygen release for adaptation to high-altitude hypoxia.
Sphingosine-1-phosphate promotes erythrocyte glycolysis and oxygen release for adaptation to high-altitude hypoxia.
Nature communications (2016-07-16)
Kaiqi Sun, Yujin Zhang, Angelo D'Alessandro, Travis Nemkov, Anren Song, Hongyu Wu, Hong Liu, Morayo Adebiyi, Aji Huang, Yuan E Wen, Mikhail V Bogdanov, Alejandro Vila, John O'Brien, Rodney E Kellems, William Dowhan, Andrew W Subudhi, Sonja Jameson-Van Houten, Colleen G Julian, Andrew T Lovering, Martin Safo, Kirk C Hansen, Robert C Roach, Yang Xia
ABSTRACT
Sphingosine-1-phosphate (S1P) is a bioactive signalling lipid highly enriched in mature erythrocytes, with unknown functions pertaining to erythrocyte physiology. Here by employing nonbiased high-throughput metabolomic profiling, we show that erythrocyte S1P levels rapidly increase in 21 healthy lowland volunteers at 5,260 m altitude on day 1 and continue increasing to 16 days with concurrently elevated erythrocyte sphingonisne kinase 1 (Sphk1) activity and haemoglobin (Hb) oxygen (O2) release capacity. Mouse genetic studies show that elevated erythrocyte Sphk1-induced S1P protects against tissue hypoxia by inducing O2 release. Mechanistically, we show that intracellular S1P promotes deoxygenated Hb anchoring to the membrane, enhances the release of membrane-bound glycolytic enzymes to the cytosol, induces glycolysis and thus the production of 2,3-bisphosphoglycerate (2,3-BPG), an erythrocyte-specific glycolytic intermediate, which facilitates O2 release. Altogether, we reveal S1P as an intracellular hypoxia-responsive biolipid promoting erythrocyte glycolysis, O2 delivery and thus new therapeutic opportunities to counteract tissue hypoxia.
Sigma-Aldrich
Sphingosine 1-phosphate, ≥95%, powder
Pricing and availability is not currently available.