Merck
  • A novel transthyretin/STAT4/miR-223-3p/FBXW7 signaling pathway affects neovascularization in diabetic retinopathy.
A novel transthyretin/STAT4/miR-223-3p/FBXW7 signaling pathway affects neovascularization in diabetic retinopathy.
Molecular and cellular endocrinology (2019-08-16)
Jun Shao, Guangming Fan, Xiaowen Yin, Yu Gu, Xiaolu Wang, Yu Xin, Yong Yao
ABSTRACT
MicroRNAs (miRNAs) are small RNAs without protein-coding functions that negatively regulate target genes and play important roles in physiological and pathological processes. The aim of this work was to reveal a novel miRNA/gene pathway in diabetic retinopathy (DR). A microarray was used to screen miRNAs in samples from nondiabetic controls and patients with DR, and miR-223-3p was screened as a potential candidate. Quantitative real-time PCR (qRT-PCR) revealed that the level of miR-223-3p was frequently overexpressed in DR samples and human retinal endothelial cells (hRECs) in hyperglycemia, but it was decreased in hyperglycemia after the addition of transthyretin (TTR). In addition, according to cell proliferation, tube formation, and wound healing assays, the downregulation of miR-223-3p suppressed cell migration and proliferation, whereas miR-223-3p upregulation showed the opposite effects. Furthermore, luciferase assays identified F-box and WD repeat domain-containing 7 (FBXW7) as a target mRNA of miR-223-3p. High glucose conditions facilitated the recruitment of signal transducer and activator of transcription 4 (STAT4) and promoted the transcription of miR-223-3p. In hRECs, in a hyperglycemic environment, TTR inhibited STAT4 expression, downregulated the level of miR-223-3p, and finally promoted FBXW7 expression. This study found a novel mechanism whereby TTR might affect neovascularization through a newly identified STAT4/miR-223-3p/FBXW7 cascade in DR.
Sigma-Aldrich
MISSION® esiRNA, targeting human FBXW7
Pricing and availability is not currently available.