Merck
  • Microwave extraction of Salvia officinalis essential oil and assessment of its GC-MS identification and protective effects versus vanadium-induced nephrotoxicity in Wistar rats models.
Microwave extraction of Salvia officinalis essential oil and assessment of its GC-MS identification and protective effects versus vanadium-induced nephrotoxicity in Wistar rats models.
Archives of physiology and biochemistry (2018-06-10)
Fatma Ghorbel Koubaa, Raed Abdennabi, Ahlem Soussi Ben Salah, Abdelfattah El Feki
ABSTRACT
In this study, we assess the impact of Salvia officinalis essential oil on renal toxicity induced by vanadium in rats. The animals were exposed to either ammonium metavanadate (5 mg/kg body weight) or the combination of vanadium and S. officinalis essential oil (15 mg EO/kg body weight) for 10 days. Vanadium induced significant renal damage, demonstrated by increased plasma levels of urea and creatinine. A marked increase in lipid peroxidation markers and carbonyl protein levels with a significant decrease in enzymatic antioxidants (SOD), catalase (CAT), and glutathione peroxidase (GPx) was also observed in vanadium-treated rats. Histopathological studies also showed vanadium-induced alterations. Concomitant administration of sage essential oil significantly restored biochemical markers and pathological lesions. This protective effect seems to be due to the richness of this extract in β-caryophyllene, limonene, carvacrol, caryophyllene, borneol and α-pinene, and α-pinene and α-thujene. These rates are determined by GC MS.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)