Merck
  • Catalpol attenuates cardiomyocyte apoptosis in diabetic cardiomyopathy via Neat1/miR-140-5p/HDAC4 axis.
Catalpol attenuates cardiomyocyte apoptosis in diabetic cardiomyopathy via Neat1/miR-140-5p/HDAC4 axis.
Biochimie (2019-05-13)
GuoLiang Zou, WeiLi Zhong, Fan Wu, XiaoXue Wang, Li Liu
ABSTRACT
The cardioprotection of catalpol and its mechanism in diabetic cardiomyopathy (DCM) remains unclear. Here, mouse cardiomyocytes were treated with high glucose (HG) to establish a model of cellular injury induced by HG. In vitro experiments were carried out and confirmed that Catalpol attenuated HG-induced long noncoding RNA (lncRNA) nuclear paraspeckle assembly transcript 1 (Neat1) expression in mouse cardiomyocytes. Mechanistically, luciferase reporter analysis indicated that Neat1 could decrease the transcription of miR-140-5p to positively regulate histone deacetylase 4 (HDAC4) expression. Notably, overexpression of miR-140-5p or silencing of HDAC4 rescued Neat1-induced cardiomyocyte apoptosis. DCM was induced in male C57BL/6 mice by intraperitoneal injection with streptozotocin (STZ) combined with a high-fat/high-sugar diet. Further in vivo experiments identified that Catalpol alleviated myocardial damage by regulating Neat1/miR-140-5p/HDAC4 axis in DCM mice. Thus, our results demonstrated that Catalpol could exert cardioprotective effect against DCM via Neat1/miR-140-5p/HDAC4 pathway.
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC4
Pricing and availability is not currently available.