Merck
  • EFFECTS OF EXENDIN-4 ON ENDOPLASMIC RETICULUM STRESS-MEDIATED INSULIN RESISTANCE IN 3T3-L1 ADIPOCYTES.
EFFECTS OF EXENDIN-4 ON ENDOPLASMIC RETICULUM STRESS-MEDIATED INSULIN RESISTANCE IN 3T3-L1 ADIPOCYTES.
Acta endocrinologica (Bucharest, Romania : 2005) (2019-09-12)
B Guan, L Wang, L Ma, X Liu, L Liu
ABSTRACT
Endoplasmic reticulum stress (ERS) is suspected as an important factor in the initiation of insulin resistance. To explore the effects of exendin-4 (Ex-4) on the endoplasmic reticulum stress (ERS)-mediated insulin resistance in 3T3-L1 adipocytes. In our study, 3T3-L1 adipocytes were pre-treated with ERS inhibitors tauroursodeoxycholic acid (TUDCA), Ex-4 and an ERS inducer tunicamycin (TM) then induced insulin resistance. Glucose consumption of the adipocytes was measured. Western blots determined the protein levels of ERS markers and insulin signaling pathway. TM treatment reduced insulin-stimulated glucose consumption by 19.7% in 3T3-L1 adipocytes. This repression was blunted by 24h pre-treatment with TUDCA or Ex-4. Ex-4 augmented insulin-stimulated glucose consumption in adipocytes by 14.9%. Western blotting showed that TM treatment significantly increased the ER stress markers including p-IRE, p-JNK, p-PERK, p-eIF2a and ATF6 expression, whereas 24h pre-treatment of adipocytes with TUDCA or Ex-4 alleviated the ER stress. Ex-4 alleviates ERS-induced insulin resistance by upregulating the expression of phosphorylated Akt. ERs mediates insulin resistance in 3T3-L1 adipocytes, and exendin-4 significantly improves this insulin resistance.