Merck
  • Redox-Induced Oxidative C-C Bond Cleavage of 2,2'-Pyridil in Diruthenium Complexes.
Redox-Induced Oxidative C-C Bond Cleavage of 2,2'-Pyridil in Diruthenium Complexes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-05-28)
Farheen Fatima Khan, Sebastian Sobottka, Biprajit Sarkar, Goutam Kumar Lahiri
ABSTRACT
In the recent years, there has been an emerging research interest in the domain of C-C bond-cleavage reactions. The present contribution deals with the redox-mediated dioxygen activation and C-C bond cleavage in a diruthenium complex [(acac)2 RuII (μ-L1)RuII (acac)2 ], 1 (acac=acetylacetonate) incorporating 2,2'-pyridil (L1) as the bridging ligand. The above process leads to a C-C-cleaved monomeric product [(acac)2 RuIII (pic- )], 2 (pic- =piconilate). Intriguingly, similar diastereomeric complexes [(acac)2 RuII (μ-L2)RuII (acac)2 ], meso (ΔΛ): 3 a and rac (ΔΔ/ΛΛ): 3 b, involving an analogous diimine bridge (L2=N1,N2-diphenyl-1,2-di(pyridin-2-yl)ethane-1,2-diimine), were stable towards such oxidative transformations. Electrochemical and spectroelectrochemical studies, in combination, establish the potential non-innocent feature of the 2,2'-Pyridil (L1) and its derivative (L2) both in oxidation and reduction processes. Additionally, theoretical calculations have been employed to verify the redox states and their behavior. Furthermore, transition state (TS) calculations at the M06L/6-31G*/LANL2DZ level of theory together with detailed kinetic studies outline a putative mechanism for the selective transformation of 1→2 involving the formation of an intermediate bearing peroxide linkage to complex 1.
Sigma-Aldrich
2,2′-Pyridil, 97%
Pricing and availability is not currently available.