Merck
  • Hypothermia induced by infusion of methionine sulfoximine into the dorsal raphe nucleus of the rat: involvement of 5-HT1A and GABAB receptors.
Hypothermia induced by infusion of methionine sulfoximine into the dorsal raphe nucleus of the rat: involvement of 5-HT1A and GABAB receptors.
European journal of pharmacology (1993-04-28)
M Ginefri-Gayet, J Gayet
ABSTRACT
L-Methionine-D,L-sulfoximine (MSO) (25 micrograms) infused locally into the dorsal raphe nucleus (DRN) induced a progressive decrease in body temperature in conscious restrained rats kept at an ambient temperature of 23 degrees C. Pretreatment with (+/-)-pindolol (3 mg/kg s.c.) significantly attenuated MSO-induced hypothermia, but pretreatment with ketanserin (0.5 mg/kg i.p.) did not alter either the magnitude or the time course of the decrease in body temperature after intra-DRN infusion of MSO. Local accumulation of gamma-aminobutyric acid (GABA) after infusion of gamma-vinylGABA (10 micrograms) and (+/-)-nipecotic acid (6 micrograms) inhibited MSO-induced hypothermia. Blockade of GABAA receptors by infusion of (-)-bicuculline (25 ng) had no effect on the decrease in body temperature elicited by MSO, but blockade of GABAB receptors by infusion of 2-OH-saclofen (13.3 ng) significantly attenuated MSO-induced hypothermia. In conclusion, local infusion of MSO into the DRN must have slowed down the rate of 5-HT turnover in serotonergic neurons and decreased the release and synthesis of GABA. 5-HT1A somato-dendritic autoreceptors and GABAB postsynaptic receptors both appear to be involved in these neurocytochemical processes leading to hypothermia.
Sigma-Aldrich
L-Methionine sulfoximine, suitable for cell culture