Merck
  • Quantification of Catalytic Activity for Electrostatically Enhanced Thioureas via Reaction Kinetics and UV-vis Spectroscopic Measurement.
Quantification of Catalytic Activity for Electrostatically Enhanced Thioureas via Reaction Kinetics and UV-vis Spectroscopic Measurement.
The Journal of organic chemistry (2018-07-20)
Yang Fan, Curtis Payne, Steven R Kass
ABSTRACT
Charged thiourea derivatives containing one and two methylated or octylated pyridinium ion centers and a tetraarylborate or triflate counteranion are reported. These novel catalysts are much more active in the Friedel-Crafts reactions of trans-β-nitroalkenes with N-methylindoles than the privileged N, N'-bis(3,5-bis(trifluoromethyl)phenyl)thiourea (i.e., Schreiner's thiourea) by up to 2-3 orders of magnitude. A previously reported UV-vis spectroscopic method by Kozlowski et al. was exploited to rationalize their reactivity order along with noncharged analogues. These results offer a new design strategy for organocatalysts by introducing positively charged centers without adding additional N-H, O-H, or S-H hydrogen bond donor sites.