Merck
  • NCα-gem-dimethylated peptoid side chains: A novel approach for structural control and peptide sequence mimetics.
NCα-gem-dimethylated peptoid side chains: A novel approach for structural control and peptide sequence mimetics.
Biopolymers (2019-03-22)
Radhe Shyam, Lionel Nauton, Gaetano Angelici, Olivier Roy, Claude Taillefumier, Sophie Faure
ABSTRACT
The design of linear peptoid oligomers adopting well-defined secondary structures while mimicking defined peptide primary sequences is a major challenge in the context of drug discovery. To this end, chemists have developed cis-inducing peptoid side chains to build robust polyproline type I helices. However, the number of efficient examples remains scarce and chemical diversity accessible through the use of these side chains is limited. Herein, we introduce an array of NCα-gem-dimethylated peptoid residues mimicking proteinogenic amino acids. Submonomer synthesis and block-coupling approaches were explored to access heterooligomers incorporating these novel types of side chains. NMR studies of monomer and trimer models showed that the NCα-gem-dimethylated groups exert complete cis control on the backbone amide conformation. Lastly, a preliminary molecular modeling study gave an insight into the preferred orientation of the substituents of the NCα-gem-dimethyl side chains relative to the peptoid backbone.