- From Antihistamine to Anti-infective: Loratadine Inhibition of Regulatory PASTA Kinases in Staphylococci Reduces Biofilm Formation and Potentiates β-Lactam Antibiotics and Vancomycin in Resistant Strains of Staphylococcus aureus.
From Antihistamine to Anti-infective: Loratadine Inhibition of Regulatory PASTA Kinases in Staphylococci Reduces Biofilm Formation and Potentiates β-Lactam Antibiotics and Vancomycin in Resistant Strains of Staphylococcus aureus.
Staphylococcus epidermidis and Staphylococcus aureus are important human pathogens responsible for two-thirds of all postsurgical infections of indwelling medical devices. Staphylococci form robust biofilms that provide a reservoir for chronic infection, and antibiotic-resistant isolates are increasingly common in both healthcare and community settings. Novel treatments that can simultaneously inhibit biofilm formation and antibiotic-resistance pathways are urgently needed to combat the increasing rates of antibiotic-resistant infections. Herein we report that loratadine, an FDA-approved antihistamine, significantly inhibits biofilm formation in both S. aureus and S. epidermidis. Furthermore, loratadine potentiates β-lactam antibiotics in methicillin-resistant strains of S. aureus and potentiates both β-lactam antibiotics and vancomycin in vancomycin-resistant strains of S. aureus. Additionally, we elucidate loratadine's mechanism of action as a novel inhibitor of the regulatory PASTA kinases Stk and Stk1 in S. epidermidis and S. aureus, respectively. Finally, we describe how Stk1 inhibition affects the expression of genes involved in both biofilm formation and antibiotic resistance in S. epidermidis and S. aureus.