Merck
  • N-arylmethylideneaminophthalimide: Design, Synthesis and Evaluation as Analgesic and Anti-inflammatory Agents.
N-arylmethylideneaminophthalimide: Design, Synthesis and Evaluation as Analgesic and Anti-inflammatory Agents.
Mini reviews in medicinal chemistry (2018-04-26)
Nasimossadat Banarouei, Asghar Davood, Hamed Shafaroodi, Ghazaleh Saeedi, Abbas Shafiee
ABSTRACT
N-aryl derivatives of phthalimide and 4-nitro phthalimide have demonstrated cyclooxygenase inhibitory activity. Also, they possess excellent analgesic and antiinflammatory activity. In this work, a new series of N-arylmethylideneamino derivatives of phthalimide and 4-nitro phthalimide were designed and synthesized. The designed compounds were synthesized by condensation of the appropriate aldehyde and N-aminophthalimide in ethanol at room temperature at PH around 3. Their analgesic and antiinflammatory activity were evaluated by acetic acid-induced pain test and carrageenan-induced paw edema test in mice and rats, respectively. The details of the synthesis and chemical characterization of the analogs are described. In vivo screening showed compounds 3a, 3b, 3f and 3h were the most potent analgesic compounds. In addition, compounds 3a, 3c, 3d, 3e and 3j indicated comparable anti-inflammatory activity to indomethacin as a reference drug.