Merck
  • Increased tau phosphorylation follows impeded dopamine clearance in a P301L and novel P301L/COMT-deleted (DM) tau mouse model.
Increased tau phosphorylation follows impeded dopamine clearance in a P301L and novel P301L/COMT-deleted (DM) tau mouse model.
Journal of neurochemistry (2018-09-22)
Jeremy Koppel, Heidy Jimenez, Leslie Adrien, Eric H Chang, Anil K Malhotra, Peter Davies
ABSTRACT
In Alzheimer's disease, the phosphorylation of tau is a critical event preceding the formation of neurofibrillary tangles. Previous work exploring the impact of a dopamine blocking antipsychotic on tau phosphorylation in a tau transgenic model suggested that extracellular dopamine may play a regulatory role in the phosphorylation state of tau. In order to test this hypothesis, and in order to develop a mouse model of impaired dopamine metabolism and tauopathy, an extant P301L transgenic tau model of Alzheimer's disease and a novel P301L/catechol-O-methyltransferase deleted model (DM mouse) were treated with the norepinephrine reuptake inhibitor reboxetine, and prefrontal dopamine concentrations and the phosphorylated state of tau was quantified. In two experiments, male and female P301L+/+//COMT+/+ and P301L+/+//COMT-/- (DM) mice were treated with reboxetine 20 mg/kg IP. In one experiment, acutely following reboxetine injection, the prefrontal cortex of mice were microdialyzed for dopamine, and its metabolites, 3,4-dihydroxyphenylacetic acid and homovanillic acid, utilizing the MetaQuant technique. In another experiment, acutely following reboxetine injections, tau phosphorylation was quantified in the frontal cortex, striatum, and hippocampus of the mice. Reboxetine injections were followed by significant increases from baseline in extracellular dopamine concentrations in P301L and DM mice, with significantly higher peak levels in the DM mice. Treatment was also followed by increases in tau phosphorylation spread throughout brain regions, with a larger impact on female mice. Extracellular dopamine concentrations exert an influence on the phosphorylation state of tau, with surges in dopamine associating with acute increases in tau phosphorylation.
Sigma-Aldrich
Reboxetine mesylate hydrate, >96% (HPLC), solid
Pricing and availability is not currently available.
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Pricing and availability is not currently available.