Merck
Targeting JNK pathway promotes human hematopoietic stem cell expansion.
Cell discovery (2019-01-10)
Xiong Xiao, Weifeng Lai, Huangfan Xie, Yang Liu, Weijie Guo, Yifang Liu, Yu Li, Yuanjun Li, Jingliang Zhang, Wenhan Chen, Minhui Shi, Lijun Shang, Ming Yin, Chengyan Wang, Hongkui Deng
ABSTRACT
The limited number of human hematopoietic stem cells (HSCs) has restrained their widespread clinical application. Despite great efforts in recent years, the in vitro expansion of HSCs remains a challenge due to incomplete understanding of the signaling networks underlying HSC self-renewal. Here, we show that culturing human cord blood (CB) CD34+ cells with JNK-IN-8, an inhibitor of the JNK signaling pathway, can enhance the self-renewal of HSCs with a 3.88-fold increase in cell number. These cultured CD34+ cells repopulated recipient mice for 21 weeks and can form secondary engraftment that lasted for more than 21 weeks. Knockdown of c-Jun, a major downstream target in the JNK pathway, promoted the expansion of hematopoietic stem and progenitor cells (HSPCs). Our findings demonstrate a critical role of the JNK pathway in regulating HSC expansion, provide new insights into HSC self-renewal mechanism, and may lead to improved clinical application of HSCs.