Merck
  • Electrostatic-Interaction-Driven Assembly of Binary Hybrids towards Fire-Safe Epoxy Resin Nanocomposites.
Electrostatic-Interaction-Driven Assembly of Binary Hybrids towards Fire-Safe Epoxy Resin Nanocomposites.
Polymers (2019-04-10)
Lu Liu, Wei Wang, Yongqian Shi, Libi Fu, Lulu Xu, Bin Yu
ABSTRACT
Manganese dioxide (MnO₂), as a promising green material, has recently attracted considerable attention of researchers from various fields. In this work, a facile method was introduced to prepare binary hybrids by fabricating three-dimensional (3D) zinc hydroxystannate (ZHS) cubes on two-dimensional (2D) MnO₂ nanosheets towards excellent flame retardancy and toxic effluent elimination of epoxy (EP) resin. Microstructural analysis confirmed that the morphologies and structures of MnO₂@ZHS binary hybrids were well characterized, implying the successful synthesis. Additionally, the morphological characterization indicated that MnO₂@ZHS binary hybrids could achieve satisfactory interfacial interaction with the EP matrix and be well dispersed in nanocomposites. Cone calorimeter test suggested that MnO₂@ZHS binary hybrids effectively suppressed the peak of heat release rate and total heat release of EP nanocomposites, performing better than MnO₂ or ZHS alone. Condensed-phase analysis revealed that MnO₂@ZHS binary hybrids could promote the char density and graphitization degree of char residues and thereby successfully retard the permeation of oxygen and flammable gases. Moreover, through the analysis of gas phase, it can be concluded that MnO₂@ZHS binary hybrids could efficiently suppress the production of toxic gases during the degradation of EP nanocomposites. This work implies that the construction of 2D/3D binary hybrids with an interfacial interaction is an effective way to fabricate high-performance flame retardants for EP.