- Rational Molecular Design Overcoming the Long Delayed Fluorescence Lifetime and Serious Efficiency Roll-Off in Blue Thermally Activated Delayed Fluorescent Devices.
Rational Molecular Design Overcoming the Long Delayed Fluorescence Lifetime and Serious Efficiency Roll-Off in Blue Thermally Activated Delayed Fluorescent Devices.
Blue thermally activated delayed fluorescent (TADF) devices with short excited-state lifetime, high reverse intersystem crossing rate, and low-efficiency roll-off were developed by managing the molecular structure of donor-acceptor-type blue emitters. Three isomers of blue TADF emitters with a diphenyltriazine acceptor and three carbazole donors were synthesized. The position of the donor moieties in the phenyl linker connecting the donor and acceptor moieties was controlled to devise compounds with a short delayed fluorescence lifetime. A blue TADF emitter with three carbazole donors at 2-, 3-, and 4- positions of a phenyl linker shortened the excited state lifetime to 4.1 μs, showed a high external quantum efficiency of 20.4 %, and low efficiency roll-off of less than 10 % at 1000 cd m-2 . Therefore, a molecular design distorting the donors by aligning them in a consecutive way is useful to resolve the issues of long delayed fluorescence lifetime and efficiency roll-off of blue TADF devices.