Merck
  • Novel microRNA, miR-sc6, modulates Schwann cell phenotype via targeting ErbB4.
Novel microRNA, miR-sc6, modulates Schwann cell phenotype via targeting ErbB4.
Experimental and therapeutic medicine (2019-04-17)
Chu Chen, Qianyan Liu, Hao Hua, Xinghui Wang, Pan Wang, Zhiming Cui, Tianmei Qian
ABSTRACT
MicroRNAs (miRNAs) are non-coding RNAs that regulate various tissues and organs, including the nervous system. Peripheral nerve injury is a common pathology of the nervous system and leads to differential expressions of a variety of miRNAs. Previously, a group of novel miRNAs have been identified in rat proximal nerve segments after sciatic nerve transection. However, the biological functions of these novel miRNAs remain undetermined. The aim of the current study was therefore to identify the function of a novel miRNA, miR-sc6, following nerve injury. Its target genes and effects on phenotypic modulation of Schwann cells were determined using a miR-sc6 mimic transfection. These observations contribute to the understanding of miRNA involvement in peripheral nerve injury and the cognition of regulatory mechanisms in peripheral nerve regeneration.
Sigma-Aldrich
Monoclonal Anti-Thy 1.1 antibody produced in mouse, clone TN-26, purified immunoglobulin, buffered aqueous solution