Merck
  • The Natural Product Butylcycloheptyl Prodiginine Binds Pre-miR-21, Inhibits Dicer-Mediated Processing of Pre-miR-21, and Blocks Cellular Proliferation.
The Natural Product Butylcycloheptyl Prodiginine Binds Pre-miR-21, Inhibits Dicer-Mediated Processing of Pre-miR-21, and Blocks Cellular Proliferation.
Cell chemical biology (2019-06-04)
Joe S Matarlo, Lauren R H Krumpe, William F Heinz, Daniel Oh, Shilpa R Shenoy, Cheryl L Thomas, Ekaterina I Goncharova, Stephen J Lockett, Barry R O'Keefe
ABSTRACT
Identification of RNA-interacting pharmacophores could provide chemical probes and, potentially, small molecules for RNA-based therapeutics. Using a high-throughput differential scanning fluorimetry assay, we identified small-molecule natural products with the capacity to bind the discrete stem-looped structure of pre-miR-21. The most potent compound identified was a prodiginine-type compound, butylcycloheptyl prodiginine (bPGN), with the ability to inhibit Dicer-mediated processing of pre-miR-21 in vitro and in cells. Time-dependent RT-qPCR, western blot, and transcriptomic analyses showed modulation of miR-21 expression and its target genes such as PDCD4 and PTEN upon treatment with bPGN, supporting on-target inhibition. Consequently, inhibition of cellular proliferation in HCT-116 colorectal cancer cells was also observed when treated with bPGN. The discovery that bPGN can bind and modulate the expression of regulatory RNAs such as miR-21 helps set the stage for further development of this class of natural product as a molecular probe or therapeutic agent against miRNA-dependent diseases.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder