Merck
  • The proteins encoded by the rbs operon of Escherichia coli: I. Overproduction, purification, characterization, and functional analysis of RbsA.
The proteins encoded by the rbs operon of Escherichia coli: I. Overproduction, purification, characterization, and functional analysis of RbsA.
Protein science : a publication of the Protein Society (1996-06-01)
C F Barroga, H Zhang, N Wajih, J H Bouyer, M A Hermodson
ABSTRACT
The nucleotide-binding component of the high-affinity ribose transport system of Escherichia coli, RbsA, was overproduced from a T7-7 expression vector, and the protein was purified. Biochemical analyses of the purified protein indicated that the ATP analogues, 5'-FSBA and 8-azido ATP, covalently labeled the protein, a reaction that was inhibited by ATP, but not by GTP or CTP. The pure protein exhibited low-level ATPase activity with a K(m) of about 140 microM. Analyses of bacterial strains carrying chromosomal deletions of rbsA and other rbs genes suggested that RbsA is important for the chemotaxis function, a surprising result that was not anticipated from previous studies. However, an inconsistency between the several results from deletion strains raises questions regarding the interpretations of the in vivo data.
Sigma-Aldrich
Reactive Red 120−Agarose, saline suspension, Type 3000-CL