Merck
  • Aurora B Inhibitor TAK-901 Synergizes with BCL-xL Inhibition by Inducing Active BAX in Cancer Cells.
Aurora B Inhibitor TAK-901 Synergizes with BCL-xL Inhibition by Inducing Active BAX in Cancer Cells.
Anticancer research (2017-02-10)
Saomi Murai, Jennifer Matuszkiewicz, Yuumi Okuzono, Hiroyuki Miya, Ron DE Jong
ABSTRACT
Aurora B kinase plays an essential role in chromosome segregation and cytokinesis, and is dysregulated in many cancer types, making it an attractive therapeutic target. TAK-901 is a potent aurora B inhibitor that showed efficacy in both in vitro and in vivo oncology models. We conducted a synthetic lethal siRNA screening to identify the genes that, when silenced, can potentiate the cell growth-inhibitory effect of TAK-901. B-cell lymphoma-extra large (BCL-xL) depletion by siRNA or chemical inhibition synergized with TAK-901 in cancer cell lines. As a mechanism of synthetic lethality, active BCL2 associated X, apoptosis regulator (BAX) was induced by TAK-901. BCL-xL protected cells from BAX-dependent apoptosis induction. Therefore, TAK-901 sensitizes cancer cells to BCL-xL inhibition. Polyploid cells induced by TAK-901 are vulnerable to BCL-xL inhibition. Our findings may have an impact on combination strategies with aurora B inhibitors in clinical studies.
Sigma-Aldrich
AZD1152-HQPA, ≥98% (HPLC)
Sigma-Aldrich
TAK-901, ≥98% (HPLC)