Merck
  • Arylsulfonylacetamides as bifunctional reagents for alkene aminoarylation.
Arylsulfonylacetamides as bifunctional reagents for alkene aminoarylation.
Science (New York, N.Y.) (2018-09-29)
Timothy M Monos, Rory C McAtee, Corey R J Stephenson
ABSTRACT
Alkene aminoarylation with a single, bifunctional reagent is a concise synthetic strategy. We report a catalytic protocol for the addition of arylsulfonylacetamides across electron-rich alkenes with complete anti-Markovnikov regioselectivity and excellent diastereoselectivity to provide 2,2-diarylethylamines. In this process, single-electron alkene oxidation enables carbon-nitrogen bond formation to provide a key benzylic radical poised for a Smiles-Truce 1,5-aryl shift. This reaction is redox-neutral, exhibits broad functional group compatibility, and occurs at room temperature with loss of sulfur dioxide. As this process is driven by visible light, uses readily available starting materials, and demonstrates convergent synthesis, it is well suited for use in a variety of synthetic endeavors.
Sigma-Aldrich
2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
[Ir(dFCF3ppy)2-(5,5′-dCF3bpy)]PF6, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
5,5′-Bis(trifluoromethyl)-2,2′-bipyridine, ≥95%
Pricing and availability is not currently available.