Merck
  • Phylogeny of the alpha and beta subunits of the dissimilatory adenosine-5'-phosphosulfate (APS) reductase from sulfate-reducing prokaryotes--origin and evolution of the dissimilatory sulfate-reduction pathway.
Phylogeny of the alpha and beta subunits of the dissimilatory adenosine-5'-phosphosulfate (APS) reductase from sulfate-reducing prokaryotes--origin and evolution of the dissimilatory sulfate-reduction pathway.
Microbiology (Reading, England) (2007-06-30)
Birte Meyer, Jan Kuever
ABSTRACT
Newly developed PCR assays were used to PCR-amplify and sequence fragments of the dissimilatory adenosine-5'-phosphosulfate (APS) reductase genes (aprBA) comprising nearly the entire gene locus (2.2-2.4 kb, equal to 92-94 % of the protein coding sequence) from 75 sulfate-reducing prokaryotes (SRP) of a taxonomically wide range. Comparative phylogenetic analysis included all determined and publicly available AprBA sequences from SRP and selected homologous sequences of sulfur-oxidizing bacteria (SOB). The almost identical AprB and AprA tree topologies indicated a shared evolutionary path for the aprBA among the investigated SRP by vertical inheritance and concomitant lateral gene transfer (LGT). The topological comparison of AprB/A- and 16S rRNA gene-based phylogenetic trees revealed novel LGT events across the SRP divisions. Compositional gene analysis confirmed Thermacetogenium phaeum to be the first validated strain affected by a recent lateral transfer of aprBA as a putative effect of long-term co-cultivation with a Thermodesulfovibrio species. Interestingly, the Apr proteins of SRP and SOB diverged into two phylogenetic lineages, with the SRP affiliated with the green sulfur bacteria, e.g. Chlorobaculum tepidum, while the Allochromatium vinosum-related sequences formed a distinct group. Analysis of genome data indicated that this phylogenetic separation is also reflected in the differing presence of the putative proteins functionally associated with Apr, QmoABC complex (quinone-interacting membrane-bound oxidoreductase) and AprM (transmembrane protein). Scenarios for the origin and evolution of the dissimilatory APS reductase are discussed within the context of the dissimilatory sulfite reductase (DsrAB) phylogeny, the appearance of QmoABC and AprM in the SRP and SOB genomes, and the geochemical setting of Archean Earth.
Sigma-Aldrich
10× REDTaq® PCR Reaction Buffer, To be used with REDTaq® DNA Polymerase
Pricing and availability is not currently available.