Merck
  • Synthesis of beta-hydroxy-alpha-amino acids with a reengineered alanine racemase.
Synthesis of beta-hydroxy-alpha-amino acids with a reengineered alanine racemase.
Bioorganic & medicinal chemistry letters (2008-09-02)
Kateryna Fesko, Lars Giger, Donald Hilvert
ABSTRACT
The Y265A mutant of alanine racemase (alrY265A) was evaluated as a catalyst for the synthesis of beta-hydroxy-alpha-amino acids. It promotes the PLP-dependent aldol condensation of glycine with a range of aromatic aldehydes. The desired products were obtained with complete stereocontrol at C(alpha) (ee>99%, D) and moderate to high selectivity at C(beta) (up to 97% de). The designed enzyme is thus similar to natural d-threonine aldolases in its substrate specificity and stereoselectivity. Moreover, its ability to utilize alanine as an alternative donor suggests an expanded scope of potential utility for the production of biologically active compounds.
Sigma-Aldrich
D-Threonine, ≥98% (TLC)
Pricing and availability is not currently available.