Merck
  • Kinetic and structural analysis of the increased affinity of enoyl-ACP (acyl-carrier protein) reductase for triclosan in the presence of NAD+.
Kinetic and structural analysis of the increased affinity of enoyl-ACP (acyl-carrier protein) reductase for triclosan in the presence of NAD+.
The Biochemical journal (2004-05-06)
Mili Kapoor, P L Swarna Mukhi, Namita Surolia, K Suguna, Avadhesha Surolia
ABSTRACT
The binding of enoyl-ACP (acyl-carrier protein) reductase from Plasmodium falciparum (PfENR) with its substrates and inhibitors has been analysed by SPR (surface plasmon resonance). The binding of the substrate analogue crotonoyl-CoA and coenzyme NADH to PfENR was monitored in real time by observing changes in response units. The binding constants determined for crotonoyl-CoA and NADH were 1.6x10(4) M(-1) and 1.9x10(4) M(-1) respectively. Triclosan, which has recently been demonstrated as a potent antimalarial agent, bound to the enzyme with a binding constant of 1.08x10(5) M(-1). However, there was a 300-fold increase in the binding constant in the presence of NAD+. The increase in the binding constant was due to a 17 times increase in the association rate constant (k(1)) from 741 M(-1) x s(-1) to 1.3x10(4) M(-1) x s(-1) and a 16 times decrease in the dissociation rate constant (k(-1)) from 6.84x10(-3) s(-1) to 4.2x10(-4) s(-1). These values are in agreement with those determined by steady-state kinetic analysis of the inhibition reaction [Kapoor, Reddy, Krishnasastry, N. Surolia and A. Surolia (2004) Biochem. J. 381, 719-724]. In SPR experiments, the binding of NAD+ to PfENR was not detected. However, a binding constant of 6.5x10(4) M(-1) was obtained in the presence of triclosan. Further support for these observations was provided by the crystal structures of the binary and ternary complexes of PfENR. Thus the dramatic enhancement in the binding affinity of both triclosan and NAD+ in the ternary complex can be explained by increased van der Waals contacts in the ternary complex, facilitated by the movement of residues 318-324 of the substrate-binding loop and the nicotinamide ring of NAD+. Interestingly, the results of the present study also provide a rationale for the increased affinity of NAD+ for the enzyme in the ternary complex.
Sigma-Aldrich
Crotonoyl coenzyme A trilithium salt, ~90% (HPLC)
Sigma-Aldrich
2-Butenoyl coenzyme A lithium salt, ≥90% (HPLC)