Merck
  • c-Jun induces mammary epithelial cellular invasion and breast cancer stem cell expansion.
c-Jun induces mammary epithelial cellular invasion and breast cancer stem cell expansion.
The Journal of biological chemistry (2010-01-08)
Xuanmao Jiao, Sanjay Katiyar, Nicole E Willmarth, Manran Liu, Xiaojing Ma, Neal Flomenberg, Michael P Lisanti, Richard G Pestell
ABSTRACT
The molecular mechanisms governing breast tumor cellular self-renewal contribute to breast cancer progression and therapeutic resistance. The ErbB2 oncogene is overexpressed in approximately 30% of human breast cancers. c-Jun, the first cellular proto-oncogene, is overexpressed in human breast cancer. However, the role of endogenous c-Jun in mammary tumor progression is unknown. Herein, transgenic mice expressing the mammary gland-targeted ErbB2 oncogene were crossed with c-jun(f/f) transgenic mice to determine the role of endogenous c-Jun in mammary tumor invasion and stem cell function. The excision of c-jun by Cre recombinase reduced cellular migration, invasion, and mammosphere formation of ErbB2-induced mammary tumors. Proteomic analysis identified a subset of secreted proteins (stem cell factor (SCF) and CCL5) induced by ErbB2 expression that were dependent upon endogenous c-Jun expression. SCF and CCL5 were identified as transcriptionally induced by c-Jun. CCL5 rescued the c-Jun-deficient breast tumor cellular invasion phenotype. SCF rescued the c-Jun-deficient mammosphere production. Endogenous c-Jun thus contributes to ErbB2-induced mammary tumor cell invasion and self-renewal.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.