Merck
  • Noradrenaline activation of neurotrophic pathways protects against neuronal amyloid toxicity.
Noradrenaline activation of neurotrophic pathways protects against neuronal amyloid toxicity.
Journal of neurochemistry (2010-02-06)
Scott E Counts, Elliott J Mufson
ABSTRACT
Degeneration of locus coeruleus (LC) noradrenergic forebrain projection neurons is an early feature of Alzheimer's disease. The physiological consequences of this phenomenon are unclear, but observations correlating LC neuron loss with increased Alzheimer's disease pathology in LC projection sites suggest that noradrenaline (NA) is neuroprotective. To investigate this hypothesis, we determined that NA protected both hNT human neuronal cultures and rat primary hippocampal neurons from amyloid-beta (Abeta(1-42) and Abeta(25-35)) toxicity. The noradrenergic co-transmitter galanin was also effective at preventing Abeta-induced cell death. NA inhibited Abeta(25-35)-mediated increases in intracellular reactive oxygen species, mitochondrial membrane depolarization, and caspase activation in hNT neurons. NA exerted its neuroprotective effects in these cells by stimulating canonical beta(1) and beta(2) adrenergic receptor signaling pathways involving the activation of cAMP response element binding protein and the induction of endogenous nerve growth factor (NGF) and brain-derived neurotrophic factor (BDNF). Treatment with functional blocking antibodies for either NGF or BDNF blocked NA's protective actions against Abeta(1-42) and Abeta(25-35) toxicity in primary hippocampal and hNT neurons, respectively. Taken together, these data suggest that the neuroprotective effects of noradrenergic LC afferents result from stimulating neurotrophic NGF and BDNF autocrine or paracrine loops via beta adrenoceptor activation of the cAMP response element binding protein pathway.
Sigma-Aldrich
Anti-Nerve Growth Factor-β Antibody, serum, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Nerve Growth Factor Antibody, clone 27/21, clone 27/21, Chemicon®, from mouse
Pricing and availability is not currently available.