Merck
The effect of photopolymerization on stem cells embedded in hydrogels.
Biomaterials (2008-10-22)
Natalja E Fedorovich, Marion H Oudshoorn, Daphne van Geemen, Wim E Hennink, Jacqueline Alblas, Wouter J A Dhert
ABSTRACT
Photopolymerizable hydrogels, formed by UV-exposure of photosensitive polymers in the presence of photoinitiators, are widely used materials in tissue engineering research employed for cellular entrapment and patterning. During photopolymerization, the entrapped cells are directly exposed to polymer and photoinitiator molecules. To develop strategies that prevent potential photoexposure-damage to osteoprogenitor cells, it is important to further characterize the effects of photopolymerization on the exposed cells. In this study we analyzed the viability, proliferation and osteogenic differentiation of multipotent stromal cell (MSC) monolayers after exposure to UV-light in the presence of Irgacure 2959, a frequently used photoinitiator in tissue engineering research. Cell cycle progression, apoptosis and osteogenic differentiation of encapsulated goat MSCs were studied in photopolymerized methacrylate-derivatized hyaluronic acid hydrogel and methacrylated hyperbranched polyglycerol gel. We demonstrate adverse effects of photopolymerization on viability, proliferation and reentry into the cell cycle of the exposed cells in monolayers, whereas the MSCs retain the ability to differentiate towards the osteogenic lineage. We further show that upon encapsulation in photopolymerizable hydrogels the viability of the embedded cells is unaffected by the photopolymerization conditions, while osteogenic differentiation depends on the type of hydrogel used.
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests
Pricing and availability is not currently available.