Merck
  • A cellular model for Friedreich Ataxia reveals small-molecule glutathione peroxidase mimetics as novel treatment strategy.
A cellular model for Friedreich Ataxia reveals small-molecule glutathione peroxidase mimetics as novel treatment strategy.
Human molecular genetics (2002-11-06)
Matthias L Jauslin, Thomas Wirth, Thomas Meier, Fabrice Schoumacher
ABSTRACT
Friedreich Ataxia (FRDA), the most prevalent of the inherited ataxias, is a multi-systemic disease with loss of sensory neurons and life-threatening hypertrophic cardiomyopathy as its most severe manifestations. Reduced levels of the mitochondrial protein frataxin lead to cell-damaging oxidative stress and consequently FRDA is considered as a model for more common neurodegenerative disorders in which reactive radicals and oxidative stress are involved. We have developed a cellular assay system that discriminates between fibroblasts from FRDA patients and unaffected donors on the basis of their sensitivity to pharmacological inhibition of de novo synthesis of glutathione. With this assay we observed that supplementation with selenium effectively improved the viability of FRDA fibroblasts, indicating that basal selenium concentrations are not sufficient to allow an adequate increase in the activity of certain detoxification enzymes (such as GPX). Furthermore, we characterized potential drug candidates and found that idebenone, a mitochondrially localized antioxidant that ameliorates cardiomyopathy in FRDA patients, as well as other lipophilic antioxidants protected FRDA cells from cell death. Our results also demonstrate for the first time that small-molecule GPX mimetics have potential as a novel treatment strategy for Friedreich Ataxia and presumably also for other neurodegenerative diseases with mitochondrial impairment.
Sigma-Aldrich
Glutathione Peroxidase Cellular Activity Assay Kit, Sufficient for 100 colorimetric tests
Pricing and availability is not currently available.