Merck
Propranolol-induced seizures in mice: the role of noradrenaline.
Cellular and molecular life sciences : CMLS (1997-08-01)
G J Amabeoku, J A Syce
ABSTRACT
The effects of some noradrenergic agents, phenobarbitone, diazepam and phenytoin on seizures produced by propranolol were investigated in mice. Isoprenaline and DL-threo-3,4-dihydroxyphenylserine (DOPS) effectively antagonized the seizures elicited by propranolol. Pargyline and imipramine significantly attenuated propranolol-induced seizures and also significantly potentiated the protecting effect of DOPS against the seizures. alpha-Methyl-p-tyrosine, disulfiram and reserpine significantly potentiated propranolol-elicited seizures. However, DOPS significantly antagonized the seizure-potentiating effects of alpha-methyl-p-tyrosine, disulfiram and reserpine. Phenylephrine, clonidine, prazosin, idazoxan, phenobarbitone, diazepam and phenytoin did not significantly alter propranolol-induced seizures. These results suggest that propranolol-induced seizures in mice may involve a noradrenergic mechanism mediated via central beta-adrenoceptors.
Sigma-Aldrich
DL-3,4-Dihydroxyphenylserine, ≥98% (HPLC), powder