Merck
  • The mitochondrial effects of small organic ligands of BCL-2: sensitization of BCL-2-overexpressing cells to apoptosis by a pyrimidine-2,4,6-trione derivative.
The mitochondrial effects of small organic ligands of BCL-2: sensitization of BCL-2-overexpressing cells to apoptosis by a pyrimidine-2,4,6-trione derivative.
The Journal of biological chemistry (2006-02-17)
Eva Milanesi, Paola Costantini, Alberto Gambalunga, Raffaele Colonna, Valeria Petronilli, Anna Cabrelle, Gianpietro Semenzato, Andrea M Cesura, Emmanuel Pinard, Paolo Bernardi
ABSTRACT
We have investigated the mitochondrial effects of BH3I-2', Chelerythrine, and HA14-1, small organic molecules that share the ability to bind the BH3 domain of BCL-2. All compounds displayed a biphasic effect on mitochondrial respiration with uncoupling at low concentrations and respiratory inhibition at higher concentrations, the relative uncoupling potency being BH3I-2' (half-maximal uncoupling at about 80 nm) > Chelerythrine (half-maximal uncoupling at about 2 microm) > HA14-1 (half-maximal uncoupling at about 20 microm). At concentrations lower than required for uncoupling all compounds sensitized the permeability transition pore (PTP) to opening both in isolated mitochondria and intact cells. To assess whether the effects on BCL-2 binding, PTP induction and respiration could be due to different structural determinants we have tested a set of HA14-1 analogs from the Hoffmann-La Roche chemical library. We have identified 5-(6-chloro-2,4-dioxo-1,3,4,10-tetrahydro-2H-9-oxa-1,3-diaza-anthracen-10-yl)-pyrimidine-2,4,6-trione (EM20-25) as a molecule devoid of effects on respiration that is able to induce PTP opening, to disrupt the BCL-2/BAX interactions in situ and to activate caspase-9 in BCL-2-overexpressing cells. EM20-25 neutralized the antiapoptotic activity of overexpressed BCL-2 toward staurosporine and sensitized BCL-2-expressing cells from leukemic patients to the killing effects of staurosporine, chlorambucil, and fludarabine. These results provide a proof of principle that the potentially toxic effects of BCL-2 ligands on mitochondrial respiration are not essential for their antiapoptotic activity and represent an important step forward in the development of tumor-selective drugs acting on BCL-2.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse