Merck
  • A Nonpump Function of Sodium Iodide Symporter in Thyroid Cancer via Cross-talk with PTEN Signaling.
A Nonpump Function of Sodium Iodide Symporter in Thyroid Cancer via Cross-talk with PTEN Signaling.
Cancer research (2018-09-16)
Fang Feng, Lamis Yehia, Ying Ni, Yi Seok Chang, Sissy Meihua Jhiang, Charis Eng
ABSTRACT
The sodium iodide symporter (NIS) is a classical iodide pump typically localized within the cell plasma membrane in thyroid cells, where NIS expression is believed to ensure success of mainstay radioiodide therapy in thyroid cancers. Although radioiodide uptake is generally reduced in thyroid cancer tissue, intracellular nonmembranous NIS has been reported to increase, suggesting that NIS serves a pump-independent function. Thyroid cancer is one of the major component cancers of Cowden syndrome, a subset of which is caused by germline mutations in PTEN In this study, we explored the noncanonical tumorigenic role of NIS in thyroid cancer cells in relation to PTEN signaling. PTEN knockdown in thyroid cancer cell lines stabilized intracellular NIS protein by promoting an interaction with NIS-LARG (leukemia-associated RhoA guanine exchange factor). Increased protein levels of cytoplasmic NIS enhanced RhoA activation and resulted in a promigration tumorigenic phenotype. Inhibition of NIS glycosylation through activation of the PI3K/AKT/mTOR signaling pathway contributed to mislocalization of NIS in the cytoplasm, facilitating its nonpump tumorigenic function through an interaction with LARG, which predominantly localized in the cytoplasm. Moreover, PTEN or PI3K/AKT/mTOR signaling could affect DPAGT1, a glycosylating enzyme involved in the initial step of N-linked glycosylation, to inhibit glycosylation of NIS. In summary, our results elucidate a pump-independent, protumorigenic role for NIS in thyroid cancer via its cross-talk with PTEN signaling.Significance: A novel pump-independent protumorigenic role of nonmembranous NIS challenges the presumption that radioiodine treatment of thyroid cancer is ineffective when transmembrane NIS is not expressed. Cancer Res; 78(21); 6121-33. ©2018 AACR.
Sigma-Aldrich
MISSION® esiRNA, targeting human DPAGT1
Pricing and availability is not currently available.
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, non-essential amino acids, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.