Merck
  • Gefitinib induces non-small cell lung cancer H1650 cell apoptosis through downregulating tumor necrosis factor-related apoptosis-inducing ligand expression levels.
Gefitinib induces non-small cell lung cancer H1650 cell apoptosis through downregulating tumor necrosis factor-related apoptosis-inducing ligand expression levels.
Oncology letters (2018-09-15)
Hanjie Yi, Shanfeng Li, Hui Li, Peng Wang, Hongyu Zheng, Xiaochun Cheng
ABSTRACT
Non-small cell lung cancer (NSCLC) presents severe threats to the lives of patients. Gefitinib is one of the first-line drugs available for the treatment of NSCLC in the clinical setting. The present study investigated the effects of gefitinib on NSCLC H1650 cell viability and apoptosis via MTT assays and flow cytometry. Western blot analysis was employed to detect tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) expression levels in H1650 cells. In the present study, H1650 cells were treated with TRAIL siRNA or an empty plasmid vector control, followed by gefitinib treatment to investigate apoptosis. Gefitinib treatment markedly inhibited H1650 cell viability, induced apoptosis and reduced TRAIL expression levels. TRAIL interference enhanced H1650 cell apoptosis induced by gefitinib. TRAIL overexpression suppressed gefitinib-induced H1650 cell apoptosis. In addition, gefitinib induced NSCLC H1650 cell apoptosis by downregulating TRAIL expression levels.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed, 0.8 mg/mL, Chemicon®
Sigma-Aldrich
Anti-beta-Actin antibody, Rabbit monoclonal, clone SP124, recombinant, expressed in proprietary host, affinity isolated antibody