Merck
  • New membrane mimetics with galactolipids: lipid properties in fast-tumbling bicelles.
New membrane mimetics with galactolipids: lipid properties in fast-tumbling bicelles.
The journal of physical chemistry. B (2013-01-11)
Weihua Ye, Jobst Liebau, Lena Mäler
ABSTRACT
Galactolipids are the main structural component of plant chloroplastic (thylakoid) membranes and of blue-green algae cell membranes. The predominant lipids in this class are monogalactosyl-diacylglycerol (MGDG) and digalactosyl-diacylglycerol (DGDG). We here present a method for the preparation of bicelles that contain these galactolipids together with a characterization of the bicelles, and the lipids within the bicelles. NMR diffusion data show that up to 30% of 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) in a q = 0.5 DMPC/DHPC lipid matrix can be replaced with either monogalactosyl-diacylglycerol or digalactosyl-diacylglycerol and that these lipids incorporate into the bicelles. No evidence for phase separation is observed. Bicelles made with monogalactosyl-diacylglycerol are significantly larger than bicelles containing only DMPC, already with only 10% of the DMPC replaced with the galactolipid. The effect of digalactosyl-diacylglycerol on bicelle size is much smaller. These observations are likely to be correlated with the different bilayer-forming properties of the lipids. Monogalactosyl-diacylglycerol is a non-bilayer-forming lipid, while digalactosyl-diacylglycerol is a bilayer-forming lipid. Both galactolipids display extensive local motion within the bilayer, as evidenced by natural abundance carbon-13 relaxation of the lipid molecules. The sugar headgroup regions are motionally restricted and cannot be described by a model that does not take into account anisotropic reorientation of the sugar units. No significant effect of the galactolipids on DMPC dynamics was observed. Our results indicate that these bicelles may become useful as model membrane mimetic media for studies of galactolipid-protein interactions.
Product Description
Avanti
MGDG, Avanti Research - A Croda Brand
Avanti
DGDG, Avanti Research - A Croda Brand