Merck
  • Transfection of chimeric anti-CD138 gene enhances natural killer cell activation and killing of multiple myeloma cells.
Transfection of chimeric anti-CD138 gene enhances natural killer cell activation and killing of multiple myeloma cells.
Molecular oncology (2014-01-07)
Hua Jiang, Wenhao Zhang, Peipei Shang, Hui Zhang, Weijun Fu, Fei Ye, Tianmei Zeng, Hejing Huang, Xueguang Zhang, Wanping Sun, Daniel Man-Yuen Sze, Qing Yi, Jian Hou
ABSTRACT
Reprogramming of NK cells with a chimeric antigen receptor (CAR) proved an effective strategy to increase NK cell reactivity and recognition specificity toward tumor cells. To enhance the cytotoxicity of NK cells against CD138-positive multiple myeloma (MM) cells, we generated genetically modified NK-92MI cells carrying a CAR that consists of an anti-CD138 single-chain variable fragment (scFv) fused to the CD3ζ chain as a signaling moiety. The genetic modification through a lentiviral vector did not affect the intrinsic cytolytic activity of NK-92MI toward human erythroleukemic cell line K562 cells or CD138-negative targets. However, these retargeted NK-92MI (NK-92MI-scFv) displayed markedly enhanced cytotoxicity against CD138-positive human MM cell lines (RPMI8226, U266 and NCI-H929) and primary MM cells at various effector-to-target ratios (E:T) as compared to the empty vector-transfected NK-92MI (NK-92MI-mock). In line with the enhanced cytotoxicity of NK-92MI-scFv, significant elevations in the secretion of granzyme B, interferon-γ and proportion of CD107a expression were also found in NK-92MI-scFv in response to CD138-positive targets compared with NK-92MI-mock. Most importantly, the enhancement in the cytotoxicity of NK-92MI-scFv did not attenuate with 10Gy-irradiation that sufficiently blocked cell proliferation. Moreover, the irradiated NK-92MI-scFv exerted definitely intensified anti-tumor activity toward CD138-positive MM cells than NK-92MI-mock in the xenograft NOD-SCID mouse model. This study provides the rationale and feasibility for adoptive immunotherapy with CD138-specific CAR-modified NK cells in CD138-positive plasmacytic malignancies, which potentially further improves remission quality and prolongs the remission duration of patients with MM after upfront chemotherapy.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution