Merck
  • Determination of Trace Antimony (III) in Water Samples with Single Drop Microextraction Using BPHA-[C4mim][PF6] System Followed by Graphite Furnace Atomic Absorption Spectrometry.
Determination of Trace Antimony (III) in Water Samples with Single Drop Microextraction Using BPHA-[C4mim][PF6] System Followed by Graphite Furnace Atomic Absorption Spectrometry.
International journal of analytical chemistry (2018-08-30)
Xiaoshan Huang, Mingxin Guan, Zhuliangzi Lu, Yiping Hang
ABSTRACT
A new sensitive method for antimony (III) determination by graphite furnace atomic absorption spectrometry (GFAAS) has been developed by using N-benzoyl-N-phenylhydroxylamine (BPHA) and 1-butyl-3-methylimidazolium hexafluorophosphate ([C4mim][PF6]) single drop microextraction. The single drop microextraction (SDMM) system is more competitive compared with other traditional extraction methods. Under the optimized conditions, the limit of detection (signal-to-noise ratio is 3) and the enrichment factor of antimony (III) are 0.01 μg·L-1 and 112, respectively. The relative standard deviation of the 0.5 μg·L-1 antimony (III) is 4.2% (n=6). The proposed method is rather sensitive to determinate trace antimony (III) in water.
Sigma-Aldrich
N-Benzoyl-N-phenylhydroxylamine, 98%
Pricing and availability is not currently available.