MilliporeSigma
  • Cloning and characterization of MST, a novel (putative) serine/threonine kinase with SH3 domain.
Cloning and characterization of MST, a novel (putative) serine/threonine kinase with SH3 domain.
Oncogene (1995-04-06)
M Katoh, M Hirai, T Sugimura, M Terada
ABSTRACT
Protein kinases play a key role in cell growth regulation. We have isolated a cDNA fragment of the MST gene from the MKN28 gastric cancer cell line cDNA pool by degenerate polymerase chain reaction. MST-cDNAs were cloned from the human brain cDNA library. Nucleotide sequence analysis indicated that the MST gene encodes a novel putative non-receptor type of serine/threonine kinase with Src homology 3 (SH3) domain, two leucine zipper domains and proline rich domain. The deduced amino acid sequence corresponding to a part of kinase domain and leucine zipper domains of MST (amino acid codons 244-461) is almost identical to the published partial amino acid sequence of MLK2. MST is the first non-receptor type of serine/threonine kinase containing SH3 domain, leucine zipper domain and proline rich domain other than PTK1/Sprk. The MST gene was moderately expressed in brain, skeletal muscle and testis as a 3.8 kb mRNA, and the MST gene has been mapped to human chromosome 19q13.1-q13.2.
Sigma-Aldrich
MLK2 (1-446), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution