Merck
Human Mob proteins regulate the NDR1 and NDR2 serine-threonine kinases.
The Journal of biological chemistry (2004-04-07)
Eric Devroe, Hediye Erdjument-Bromage, Paul Tempst, Pamela A Silver
ABSTRACT
Human NDR1 (nuclear Dbf2-related) is a widely expressed nuclear serine-threonine kinase that has been implicated in cell proliferation and/or tumor progression. Here we present molecular characterization of the human NDR2 serine-threonine kinase, which shares approximately 87% sequence identity with NDR1. NDR2 is expressed in most human tissues with the highest expression in the thymus. In contrast to NDR1, NDR2 is excluded from the nucleus and exhibits a punctate cytoplasmic distribution. The differential localization of NDR1 and NDR2 suggests that each kinase may serve distinct functions. Thus, to identify proteins that interact with NDR1 or NDR2, epitope-tagged kinases were immunoprecipitated from Jurkat T-cells. Two uncharacterized proteins that are homologous to the Saccharomyces cerevisiae kinase regulators Mob1 and Mob2 were identified. We demonstrate that NDR1 and NDR2 partially colocalize with human Mob2 in HeLa cells and confirm the NDR-Mob interactions in cell extracts. Interestingly, NDR1 and NDR2 form stable complexes with Mob2, and this association dramatically stimulates NDR1 and NDR2 catalytic activity. In summary, this work identifies a unique class of human kinase-activating subunits that may be functionally analagous to cyclins.