Merck
  • Synthesis of biologically active peptide nucleic acid-peptide conjugates by sortase-mediated ligation.
Synthesis of biologically active peptide nucleic acid-peptide conjugates by sortase-mediated ligation.
The Journal of organic chemistry (2007-04-17)
Stephan Pritz, Yvonne Wolf, Oliver Kraetke, Jana Klose, Michael Bienert, Michael Beyermann
ABSTRACT
Sortase A is a transpeptidase that cleaves at a pentapeptide-motif and subsequently transfers the acyl component to a nucleophile containing N-terminal oligoglycines. We investigate the reaction conditions of the sortase-mediated ligation and demonstrate a useful application by the synthesis of a peptide nucleic acid-cell-penetrating peptide chimera, the reaction equilibrium of which can be shifted in favor of the product by dialyzing out the low molecular weight byproduct. The synthesized conjugate exhibits dose-dependent antisense activity.
Product Description
SAFC
Sortase A, S. aureus, His-tag, Staphylococcus aureus, recombinant, N-terminal His-Tag