Merck
  • Precursor-directed fungal generation of novel halogenated chaetoglobosins with more preferable immunosuppressive action.
Precursor-directed fungal generation of novel halogenated chaetoglobosins with more preferable immunosuppressive action.
Chemical communications (Cambridge, England) (2010-12-15)
Hui Ming Ge, Wei Yan, Zhi Kai Guo, Qiong Luo, Rui Feng, Le Yun Zang, Yan Shen, Rui Hua Jiao, Qiang Xu, Ren Xiang Tan
ABSTRACT
Precursor-fed cultivation of endophytic Chaetomium globosum 1C51 afforded nine novel "unnatural" halogenated chaetoglobosins including those with more preferable immunosuppressive activity.
Sigma-Aldrich
Chaetoglobosin A, from Chaetomium globosum, ≥98% (HPLC)