Merck
  • Lupeol alters ER stress-signaling pathway by downregulating ABCG2 expression to induce Oxaliplatin-resistant LoVo colorectal cancer cell apoptosis.
Lupeol alters ER stress-signaling pathway by downregulating ABCG2 expression to induce Oxaliplatin-resistant LoVo colorectal cancer cell apoptosis.
Environmental toxicology (2018-02-13)
Ming-Cheng Chen, Hsi-Hsien Hsu, Yuan-Yuan Chu, Sue-Fei Cheng, Chia-Yao Shen, Yi-Jiun Lin, Ray-Jade Chen, Vijaya Padma Viswanadha, Yueh-Min Lin, Chih-Yang Huang
ABSTRACT
Colorectal cancer (CRC) is one of the most common cancers and causes of cancer-related death. There are several first-line chemotherapeutic drugs used to treat CRC. Oxaliplatin (OXA) is an alkylating cytotoxic agent that is usually combined with other chemotherapeutic drugs to treat stage II and stage III CRC. However, cancer cells commonly acquire multidrug resistance (MDR), which is a major obstruction to cancer treatment. Recent studies have shown that natural components from traditional Chinese medicine or foods that have many biological functions may be new adjuvant therapies in clinical trials. We challenged LoVo CRC cell lines with OXA in a dose-dependent manner to create an OXA-resistant model. The expression of ABCG2 was significantly higher, and levels of endoplasmic reticulum (ER) stress markers were lower than those Parental cells. However, Lupeol, which is found in fruits and vegetables, has been shown to have bioactive properties, including anti-tumor properties that are relevant to many diseases. In our study, Lupeol downregulated cell viability and activated cell apoptosis. Moreover, Lupeol decreased the expression of ABCG2 and activated ER stress to induce OXA-resistant cell death. Importantly, the anti-tumor effect of Lupeol in OXA-resistant cells was higher than that of LoVo Parental cells. In addition, we also confirmed our results with a xenograft animal model, and the tumor size significantly decreased after Lupeol injections. Our findings show that Lupeol served as a strong chemoresistant sensitizer and could be a new adjuvant therapy method for chemoresistant patients.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Pricing and availability is not currently available.