Merck
  • Substantial changes of cellular iron homeostasis during megakaryocytic differentiation of K562 cells.
Substantial changes of cellular iron homeostasis during megakaryocytic differentiation of K562 cells.
Development, growth & differentiation (2009-08-01)
Krzysztof Wandzik, Claudia Zahn, Katrin Dassler, Hendrik Fuchs
ABSTRACT
We investigated the remodeling of iron metabolism during megakaryocytic development of K562 cells. Differentiation was successfully verified by increase of the megakaryocytic marker CD61 and concomitant decrease of the erythroid marker γ-globin. The reduction of erythroid properties was accompanied by changes in the cellular iron content and in the expression of proteins regulating cellular iron homeostasis. Independent of available inorganic or transferrin-bound extracellular iron, total intracellular iron increases while the iron-to-protein ratio decreases. The iron exporter ferroportin is downregulated within 1-6 h, followed by downregulation of transferrin receptor-1 (TfR1) and ferritin heavy chain (H-ferritin) mainly after 24-48 h. The hemochromatosis protein-1, a ligand of TfR1, peaked after 24 h. All effects were independent of iron supply with the exception of H-ferritin, which was restored by excess iron. While alterations of CD61, TfR1 and ferritin expression were revoked by a protein kinase C inhibitor, downregulation of ferroportin remained unaffected.
Sigma-Aldrich
Anti-Mouse IgM (μ-chain specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution