Merck
  • Electronically tunable anion-π interactions in pyrylium complexes: experimental and theoretical studies.
Electronically tunable anion-π interactions in pyrylium complexes: experimental and theoretical studies.
Physical chemistry chemical physics : PCCP (2014-07-30)
Antonio Franconetti, Lidia Contreras-Bernal, Sorel Jatunov, Manuel Gómez-Guillén, Manuel Angulo, Rafael Prado-Gotor, Francisca Cabrera-Escribano
ABSTRACT
Noncovalent interactions of anions with electron-deficient aromatic rings that have been studied so far involve non-heteroaromatic or nitrogen-based heteroaromatic systems. Here we report the first case of an organic oxygenated aromatic system, in particular the tri-aryl-pyrylium tetrafluoroborate system, for which noncovalent anion-π interactions of the pyrylium cation with the tetrafluoroborate anion have been experimentally detected and demonstrated by means of (19)F NMR spectroscopy in solution. A series of pyrylium tetrafluoroborate salts were synthesized in the presence of BF3·Et2O, by direct reaction of 4-substituted benzaldehydes with 4-substituted acetophenones or via the previously obtained chalcone of the less reactive ketone. Correlations of (19)F NMR chemical shifts of tetrafluoroborate anion for most of the synthesized tri-arylpyrylium tetrafluoroborate complexes with both the pyrylium cation molecular weight and the standard substituent Hammett constants, demonstrate anion-π(+) interaction to act between the polyatomic anion BF4(-) and the pyrylium aromatic system. DFT calculations reveal that an additional (C-H)(+)-anion hydrogen bond involving the H(5) of pyrylium ring exists for these fluorescent dyes that show a tunable cup-to-cap shape cavity. The strong fluorescence emission observed for some representative pyrylium tetrafluoroborates described herein, makes them a promising class of tunable emission wavelength dyes for laser technology applications.
Sigma-Aldrich
2,4,6-Tri-(4-fluorophenyl)pyrylium tetrafluoroborate, ≥95%