Merck
  • Takinib, a Selective TAK1 Inhibitor, Broadens the Therapeutic Efficacy of TNF-α Inhibition for Cancer and Autoimmune Disease.
Takinib, a Selective TAK1 Inhibitor, Broadens the Therapeutic Efficacy of TNF-α Inhibition for Cancer and Autoimmune Disease.
Cell chemical biology (2017-08-19)
Juliane Totzke, Deepak Gurbani, Rene Raphemot, Philip F Hughes, Khaldon Bodoor, David A Carlson, David R Loiselle, Asim K Bera, Liesl S Eibschutz, Marisha M Perkins, Amber L Eubanks, Phillip L Campbell, David A Fox, Kenneth D Westover, Timothy A J Haystead, Emily R Derbyshire
ABSTRACT
Tumor necrosis factor alpha (TNF-α) has both positive and negative roles in human disease. In certain cancers, TNF-α is infused locally to promote tumor regression, but dose-limiting inflammatory effects limit broader utility. In autoimmune disease, anti-TNF-α antibodies control inflammation in most patients, but these benefits are offset during chronic treatment. TAK1 acts as a key mediator between survival and cell death in TNF-α-mediated signaling. Here, we describe Takinib, a potent and selective TAK1 inhibitor that induces apoptosis following TNF-α stimulation in cell models of rheumatoid arthritis and metastatic breast cancer. We demonstrate that Takinib is an inhibitor of autophosphorylated and non-phosphorylated TAK1 that binds within the ATP-binding pocket and inhibits by slowing down the rate-limiting step of TAK1 activation. Overall, Takinib is an attractive starting point for the development of inhibitors that sensitize cells to TNF-α-induced cell death, with general implications for cancer and autoimmune disease treatment.
Sigma-Aldrich
NG25 trihydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Takinib, ≥98% (HPLC)
Pricing and availability is not currently available.